The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000498.3 | NP_000489.3 | 7 - 8 | 1203 | 110 |
GenBank mRNA | X54741.1 | - | 7 - 8 | 1203 | 110 |