The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000847.4 | NP_000838.3 | 5 - 6 | 495 | 142 |
XM_006715072.3 | XP_006715135.1 | 3 - 4 | 329 | 142 | |
XM_011514533.2 | XP_011512835.1 | 2 - 3 | 321 | 142 | |
XM_017010794.1 | XP_016866283.1 | 3 - 4 | 370 | 142 | |
GenBank mRNA | AA846513.1 | - | 2 - 3 | 142 | |
AF020919.1 | - | 4 - 5 | 355 | 142 | |
AF508266.1 | - | 4 - 5 | 416 | 142 | |
BC020619.1 | - | 5 - 6 | 482 | 142 |