Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D10C
Entrez Gene ID
374403
Gene Name
TBC1 domain family member 10C
Gene Aliases
CARABIN, EPI64C
Chromosome Location
Chr.11: 67403718 - 67410090 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256508.1 NP_001243437.1 6 - 7 619 146
NM_198517.3 NP_940919.1 6 - 7 640 146
NR_046266.1 5 - 6 810 146
XM_006718538.3 XP_006718601.1 6 - 7 899 146
XM_006718539.3 XP_006718602.1 6 - 7 882 146
XM_006718540.3 XP_006718603.1 6 - 7 812 146
XM_006718541.3 XP_006718604.1 6 - 7 903 146
XM_006718542.3 XP_006718605.1 6 - 7 886 146
XM_006718543.2 XP_006718606.1 6 - 7 620 146
XM_006718544.3 XP_006718607.1 6 - 7 814 146
XM_011545001.2 XP_011543303.1 5 - 6 841 146
XM_011545002.2 XP_011543304.1 5 - 6 860 146
GenBank mRNA BC022202.1 - 5 - 6 584 146
BC036873.1 - 5 - 6 595 146
BC062999.1 - 6 - 7 612 146

View Full Product Details