The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079691.1 | NP_001073159.1 | 1 - 1 | 215 | 130 |
NM_001286459.1 | NP_001273388.1 | 1 - 1 | 215 | 130 | |
NM_001286460.1 | NP_001273389.1 | 1 - 1 | 215 | 130 | |
NM_001286461.1 | NP_001273390.1 | 1 - 1 | 215 | 130 | |
NM_052818.2 | NP_438169.2 | 1 - 1 | 215 | 130 | |
XM_006719890.3 | XP_006719953.1 | 1 - 1 | 471 | 130 | |
XM_011535302.2 | XP_011533604.1 | 1 - 1 | 167 | 130 | |
XM_017020836.1 | XP_016876325.1 | 1 - 1 | 1670 | 130 | |
XM_017020837.1 | XP_016876326.1 | 1 - 1 | 455 | 130 | |
XM_017020839.1 | XP_016876328.1 | 1 - 1 | 1686 | 130 | |
XM_017020840.1 | XP_016876329.1 | 1 - 1 | 194 | 130 | |
XM_017020842.1 | XP_016876331.1 | 1 - 1 | 167 | 130 | |
GenBank mRNA | AL049785.1 | - | 1 - 1 | 171 | 131 |
AL049786.1 | - | 1 - 1 | 171 | 131 | |
AL049787.1 | - | 1 - 1 | 171 | 131 | |
BC022188.2 | - | 1 - 1 | 161 | 130 | |
BI551105.1 | - | 1 - 1 | 220 | 130 | |
U50527.1 | - | 1 - 1 | 166 | 126 |