The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282419.2 | NP_001269348.1 | 4 - 4 | 1434 | 110 |
NM_001282420.2 | NP_001269349.1 | 5 - 5 | 1496 | 110 | |
NM_001282421.2 | NP_001269350.1 | 5 - 5 | 1487 | 110 | |
NM_001282422.2 | NP_001269351.1 | 4 - 4 | 1362 | 110 | |
NM_001291304.1 | NP_001278233.1 | 7 - 7 | 1673 | 110 | |
NM_005000.4 | NP_004991.1 | 5 - 5 | 1479 | 110 | |
NR_104168.2 | 6 - 6 | 1688 | 110 | ||
NR_104169.2 | 6 - 6 | 1639 | 110 | ||
NR_111925.1 | 5 - 5 | 1477 | 110 | ||
NR_111926.1 | 8 - 8 | 1637 | 110 | ||
GenBank mRNA | BC000813.1 | - | 5 - 5 | 1395 | 110 |
BC020821.1 | - | 5 - 5 | 1379 | 110 | |
BC070236.1 | - | 5 - 5 | 1380 | 110 | |
BC070237.1 | - | 5 - 5 | 1379 | 110 | |
CR933664.1 | - | 5 - 5 | 1496 | 110 | |
U53468.1 | - | 5 - 5 | 1393 | 110 | |
U64028.1 | - | 5 - 5 | 1386 | 110 |