The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153015.2 | NP_694560.1 | 1 - 2 | 135 | 84 |
GenBank mRNA | AK055230.1 | - | 1 - 2 | 61 | 84 |
BC030710.2 | - | 1 - 2 | 135 | 84 |