Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC83
Entrez Gene ID
220047
Gene Name
coiled-coil domain containing 83
Gene Aliases
CT148, HSD9
Chromosome Location
Chr.11: 85855066 - 85920021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173556.4 NP_775827.2 8 - 9 1310 71
XM_011544839.2 XP_011543141.1 8 - 9 896 71
XM_011544840.2 XP_011543142.1 8 - 9 968 71
XM_011544841.1 XP_011543143.1 8 - 9 858 71
XM_011544842.1 XP_011543144.1 8 - 9 1310 71
XM_011544843.1 XP_011543145.1 7 - 8 1142 71
XM_011544844.2 XP_011543146.1 6 - 7 1022 71
GenBank mRNA AK124113.1 - 8 - 9 1311 71

View Full Product Details