Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KAAG1
Entrez Gene ID
353219
Gene Name
kidney associated antigen 1
Gene Aliases
RU2AS
Chromosome Location
Chr.6: 24356903 - 24358284 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181337.3 NP_851854.1 1 - 1 1255 96
GenBank mRNA AF181722.1 - 1 - 1 1255 96

View Full Product Details