Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FKBP9P1
- Entrez Gene ID
-
360132
- Gene Name
- FK506 binding protein 9 pseudogene 1
- Gene Aliases
-
FKBP9L
- Chromosome Location
-
Chr.7: 55681074 - 55704567 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027342.1 |
|
1 - 2 |
392 |
84 |
GenBank mRNA |
BC011872.2 |
- |
1 - 2 |
392 |
84 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BE298844.1 |
- |
1 - 2 |
374 |
|
Chr. 7 - 55685202 - 55689283 [-] on Build GRCh38 |
84 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
vesicular, overexpressed in cancer, prosurvival protein 1 |
VOPP1 |
ECOP, GASP |
-114.4kb
|
Sense |
Chr. - 55434406 - 55572520 on Build GRCh38 |
Downstream |
putative uncharacterized protein FLJ44672 |
LOC101060341 |
|
57.2kb
|
Sense |
Chr. - 55746453 - 55750715 on Build GRCh38 |
View Full Product Details