Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWC2
Entrez Gene ID
375567
Gene Name
von Willebrand factor C domain containing 2
Gene Aliases
PSST739, UNQ739
Chromosome Location
Chr.7: 49773661 - 49923777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198570.4 NP_940972.2 2 - 3 1250 76
NR_136188.1 1 - 2 797 76
GenBank mRNA AB292671.1 - 1 - 2 694 76
AY358393.1 - 2 - 3 1274 76
BC110857.1 - 2 - 3 1193 76

View Full Product Details