The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207352.3 | NP_997235.3 | 2 - 3 | 628 | 104 |
XM_005262935.3 | XP_005262992.1 | 2 - 3 | 413 | 104 | |
GenBank mRNA | AK122600.1 | - | 2 - 3 | 363 | 104 |
AK126473.1 | - | 3 - 4 | 297 | 104 | |
AY422002.1 | - | 2 - 3 | 628 | 104 | |
BC060857.1 | - | 2 - 3 | 494 | 104 |