The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207412.2 | NP_997295.1 | 1 - 2 | 342 | 57 |
GenBank mRNA | AK125570.1 | - | 1 - 2 | 342 | 57 |
BC137511.1 | - | 1 - 2 | 337 | 57 | |
BC137512.1 | - | 1 - 2 | 337 | 57 |