Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCRP2
Entrez Gene ID
400892
Gene Name
breakpoint cluster region pseudogene 2
Gene Aliases
BCR-2, BCR2, BCRL2
Chromosome Location
Chr.22: 21103016 - 21122286 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_037566.1 3 - 4 422 110
GenBank mRNA AK124943.1 - 3 - 4 422 110

View Full Product Details