The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006827.5 | NP_006818.3 | 5 - 5 | 2523 | 137 |
GenBank mRNA | AK023615.1 | - | 1 - 1 | 321 | 137 |
AK056156.1 | - | 1 - 1 | 968 | 137 | |
AL832012.2 | - | 5 - 5 | 2467 | 137 |