Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ZSCAN23
- Entrez Gene ID
-
222696
- Gene Name
- zinc finger and SCAN domain containing 23
- Gene Aliases
-
ZNF390, ZNF453, dJ29K1.3, dJ29K1.3.1
- Chromosome Location
-
Chr.6: 28425516 - 28443554 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK092117.1 |
- |
2 - 3 |
569 |
72 |
View Full Product Details