Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPR39
Entrez Gene ID
2863
Gene Name
G protein-coupled receptor 39
Gene Aliases
-
Chromosome Location
Chr.2: 132415972 - 132646596 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK123759.1 - 1 - 2 476 118

View Full Product Details