Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DEFA9P
- Entrez Gene ID
-
449492
- Gene Name
- defensin alpha 9, pseudogene
- Gene Aliases
-
DEFAP2
- Chromosome Location
-
Chr.8: 6959289 - 6960161 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073408.1 |
|
1 - 2 |
173 |
132 |
GenBank mRNA |
AY746434.1 |
- |
1 - 2 |
173 |
132 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
defensin alpha 8, pseudogene |
DEFA8P |
DEFAP1 |
-7.7kb
|
Sense |
Chr. - 6950726 - 6951599 on Build GRCh38 |
Downstream |
defensin alpha 10, pseudogene |
DEFA10P |
|
8.0kb
|
Sense |
Chr. - 6968141 - 6969113 on Build GRCh38 |
View Full Product Details