The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146041.1 | NP_001139513.1 | 1 - 1 | 758 | 111 |
GenBank mRNA | AJ406941.1 | - | 1 - 1 | 703 | 111 |