Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IGKC
- Entrez Gene ID
-
3514
- Gene Name
- immunoglobulin kappa constant
- Gene Aliases
-
HCAK1, IGKCD, Km
- Chromosome Location
-
Chr.2: 88857361 - 88857683 on Build GRCh38
- Gene Symbol
- IGK
- Entrez Gene ID
-
50802
- Gene Name
- immunoglobulin kappa locus
- Gene Aliases
-
IGK@
- Chromosome Location
-
Chr.2: 88857361 - 90235368 on Build GRCh38
- Gene Symbol
- IGKV1-5
- Entrez Gene ID
-
28299
- Gene Name
- immunoglobulin kappa variable 1-5
- Gene Aliases
-
IGKV, IGKV15, L12, L12a, V1
- Chromosome Location
-
Chr.2: 88947301 - 88947776 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC030814.1 |
- |
4 - 4 |
772 |
109 |
BC034142.1 |
- |
4 - 4 |
775 |
109 |
BC034146.1 |
- |
4 - 4 |
758 |
109 |
BC073792.1 |
- |
4 - 4 |
743 |
109 |
- Gene Symbol
- IGKV2-24
- Entrez Gene ID
-
28923
- Gene Name
- immunoglobulin kappa variable 2-24
- Gene Aliases
-
A23, IGKV224
- Chromosome Location
-
Chr.2: 89176328 - 89177130 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC063599.1 |
- |
4 - 4 |
756 |
109 |
View Full Product Details