The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035511.1 | NP_001030588.1 | 5 - 5 | 1417 | 86 |
NM_001035512.1 | NP_001030589.1 | 5 - 5 | 1479 | 86 | |
NM_001035513.1 | NP_001030590.1 | 4 - 4 | 1422 | 86 | |
NM_001278172.1 | NP_001265101.1 | 4 - 4 | 1315 | 86 | |
NM_003001.3 | NP_002992.1 | 6 - 6 | 1581 | 86 | |
NR_103459.1 | 7 - 7 | 1608 | 86 |