Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
FKRP | NM_001039885.2 | NP_001034974.1 | ||
NM_024301.4 | NP_077277.1 | |||
XM_005259247.2 | XP_005259304.1 | |||
XM_005259248.2 | XP_005259305.1 | |||
XM_005259249.4 | XP_005259306.1 | |||
XM_011527306.1 | XP_011525608.1 | |||
XM_011527307.1 | XP_011525609.1 | |||
XM_017027297.1 | XP_016882786.1 | |||
AJ314847.1 | CAC85633.1 | |||
AK022638.1 | BAB14146.1 | |||
AK095497.1 | ||||
AK291282.1 | ||||
BM979509.1 | ||||
DA123579.1 | ||||
DB090073.1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
STRN4 | NM_001039877.1 | 1 | 255 | NP_001034966.1 |
NM_013403.2 | 1 | 255 | NP_037535.2 | |
XM_006723171.2 | 1 | 267 | XP_006723234.1 | |
XM_006723172.2 | 1 | 267 | XP_006723235.1 | |
XM_011526878.1 | 1 | 267 | XP_011525180.1 | |
XM_017026719.1 | 1 | 267 | XP_016882208.1 | |
AB209003.1 | 1 | 207 | BAD92240.1 | |
AF212940.1 | 1 | 222 | AAF29527.1 | |
AK297637.1 | ||||
AK298804.1 | ||||
BC034604.1 | 1 | 232 | AAH34604.1 | |
DA510367.1 | 1 | 255 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833850 | Chr19:46703285 - 46867053 on Build GRCh38 | Loss | SNAR-E FKRP STRN4 ARHGAP35 LOC107987269 PRKD2 SLC1A5 AP2S1 MIR320E |