The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014872.2 | NP_055687.1 | 2 - 2 | 4567 | 127 |
XM_005251634.1 | XP_005251691.1 | 2 - 2 | 5411 | 127 | |
GenBank mRNA | AB002352.1 | - | 2 - 2 | 4560 | 127 |