Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ZNF43
- Entrez Gene ID
-
7594
- Gene Name
- zinc finger protein 43
- Gene Aliases
-
HTF6, KOX27, ZNF39L1
- Chromosome Location
-
Chr.19: 21804946 - 21852095 on Build GRCh38
View Full Product Details