The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080826.1 | NP_543016.1 | 6 - 6 | 1500 | 173 |
GenBank mRNA | BC017997.1 | - | 1 - 1 | 556 | 173 |