Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BC038203.1 |
- |
1 |
824 |
|
Chr. X - 135318560 - 135319718 [-] on Build GRCh38 |
158 |
GenBankmRNA |
AI401101.1 |
- |
1 |
|
|
Chr. X - 135318560 - 135319040 [+] on Build GRCh38 |
158 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cancer/testis antigen 55 |
CT55 |
BJHCC20A, CXorf48 |
-146.7kb
|
Anti-sense |
Chr. - 135156536 - 135171827 on Build GRCh38 |
Downstream |
zinc finger protein 449 |
ZNF449 |
ZSCAN19 |
25.7kb
|
Sense |
Chr. - 135344098 - 135363413 on Build GRCh38 |
View Full Product Details