The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001547.4 | NP_001538.4 | 2 - 2 | 2859 | 142 |
GenBank mRNA | BC032839.2 | - | 2 - 2 | 2762 | 142 |