The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001663.3 | NP_001654.1 | 2 - 2 | 1859 | 148 |
GenBank mRNA | AF047432.1 | - | 1 - 1 | 1313 | 148 |
BC030291.1 | - | 1 - 1 | 568 | 148 |