Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOXI2
Entrez Gene ID
399823
Gene Name
forkhead box I2
Gene Aliases
-
Chromosome Location
Chr.10: 127737274 - 127741186 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207426.2 NP_997309.2 2 - 2 3026 94
GenBank mRNA AK128865.1 - 2 - 2 2626 94

View Full Product Details