Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR5M1
Entrez Gene ID
390168
Gene Name
olfactory receptor family 5 subfamily M member 1
Gene Aliases
OR11-208, OST050
Chromosome Location
Chr.11: 56612555 - 56613502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004740.1 NP_001004740.1 1 - 1 363 146

View Full Product Details