Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR52L1
Entrez Gene ID
338751
Gene Name
olfactory receptor family 52 subfamily L member 1
Gene Aliases
OR11-50
Chromosome Location
Chr.11: 5985892 - 5986985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005173.3 NP_001005173.3 1 - 1 1008 73
GenBank mRNA BC137343.1 - 1 - 1 1008 73

View Full Product Details