Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POTEA
Entrez Gene ID
340441
Gene Name
POTE ankyrin domain family member A
Gene Aliases
A26A1, CT104.3, POTE-8, POTE8
Chromosome Location
Chr.8: 43292370 - 43363185 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002920.1 NP_001002920.1 11 - 12 1483 112
NM_001005365.2 NP_001005365.2 12 - 13 1621 112
GenBank mRNA AY462869.1 - 12 - 13 1621 112
AY462870.1 - 11 - 12 1483 112

View Full Product Details