Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DALRD3
Entrez Gene ID
55152
Gene Name
DALR anticodon binding domain containing 3
Gene Aliases
-
Chromosome Location
Chr.3: 49015399 - 49021386 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009996.2 NP_001009996.1 6 - 7 1040 168
NM_001276405.1 NP_001263334.1 6 - 7 1040 168
NM_018114.5 NP_060584.3 6 - 7 1057 168
XM_005265269.3 XP_005265326.1 6 - 7 1021 168
XM_006713219.2 XP_006713282.1 7 - 8 1176 168
XM_011533891.1 XP_011532193.1 6 - 7 1021 168
XM_011533892.1 XP_011532194.1 6 - 7 1372 168
XM_017006723.1 XP_016862212.1 7 - 8 1176 168
GenBank mRNA AK001358.1 - 5 - 6 1025 168
AK093204.1 - 6 - 7 1021 168
AK093294.1 - 5 - 6 828 168
AK301352.1 - 6 - 7 1008 168
AK308273.1 - 7 - 8 1134 168
BC014099.2 - 5 - 6 1079 168
BC032440.1 - 6 - 7 1003 168
BC047683.1 - 6 - 7 1002 168
BC054493.1 - 6 - 7 996 168

View Full Product Details