The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033514.4 | NP_277049.1 | 2 - 3 | 641 | 141 |
GenBank mRNA | AF288404.1 | - | 2 - 3 | 381 | 141 |
BC093812.1 | - | 2 - 3 | 366 | 141 | |
BC112233.1 | - | 2 - 3 | 366 | 141 | |
BX108353.1 | - | 2 - 3 | 366 | 141 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205288.1 | NP_001192217.1 | 2 - 3 | 641 | 141 |