The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177316.1 | NP_001170787.1 | 10 - 11 | 1316 | 173 |
NM_001177317.1 | NP_001170788.1 | 10 - 11 | 1319 | 173 | |
NM_080877.2 | NP_543153.1 | 10 - 11 | 1281 | 173 | |
XM_011518257.2 | XP_011516559.1 | 10 - 11 | 1257 | 173 | |
XM_011518258.1 | XP_011516560.1 | 9 - 10 | 1147 | 173 | |
XM_011518259.1 | XP_011516561.1 | 10 - 11 | 1281 | 173 | |
XM_011518260.1 | XP_011516562.1 | 10 - 11 | 1199 | 173 | |
XM_011518261.2 | XP_011516563.1 | 10 - 11 | 1272 | 173 | |
XM_017014290.1 | XP_016869779.1 | 11 - 12 | 1367 | 173 | |
XM_017014291.1 | XP_016869780.1 | 10 - 11 | 1257 | 173 | |
XM_017014292.1 | XP_016869781.1 | 10 - 11 | 1257 | 173 | |
GenBank mRNA | AB055000.1 | - | 10 - 11 | 1165 | 173 |
AK095999.1 | - | 10 - 11 | 1281 | 173 |