Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEPHL1
Entrez Gene ID
341208
Gene Name
hephaestin like 1
Gene Aliases
-
Chromosome Location
Chr.11: 94021212 - 94114208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098672.1 NP_001092142.1 17 - 18 3189 111
GenBank mRNA BX641008.1 - 6 - 7 944 111

View Full Product Details