The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024455.3 | NP_001019626.1 | 1 - 2 | 3735 | 69 |
GenBank mRNA | AB082532.1 | - | 1 - 2 | 3735 | 69 |
AL359579.2 | - | 1 - 2 | 2779 | 69 | |
BC151224.1 | - | 1 - 2 | 3756 | 69 |