Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPIP5K1
Entrez Gene ID
9677
Gene Name
diphosphoinositol pentakisphosphate kinase 1
Gene Aliases
HISPPD2A, IP6K, IPS1, VIP1, hsVIP1
Chromosome Location
Chr.15: 43533462 - 43590272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130858.2 NP_001124330.1 14 - 15 1641 117
NM_001130859.2 NP_001124331.1 13 - 14 1636 117
NM_001190214.1 NP_001177143.1 14 - 15 1641 117
NM_014659.5 NP_055474.3 13 - 14 1559 117
XM_005254803.1 XP_005254860.1 14 - 15 1641 117
XM_005254804.1 XP_005254861.1 14 - 15 1641 117
XM_006720786.2 XP_006720849.1 14 - 15 1641 117
XM_011522249.1 XP_011520551.1 14 - 15 1641 117
XM_011522250.1 XP_011520552.1 14 - 15 1744 117
XM_011522252.1 XP_011520554.1 13 - 14 1615 117
XM_011522254.1 XP_011520556.1 13 - 14 1511 117
XM_011522258.1 XP_011520560.1 8 - 9 901 117
XM_011522259.1 XP_011520561.1 14 - 15 1641 117
XM_017022735.1 XP_016878224.1 13 - 14 1958 117
XM_017022736.1 XP_016878225.1 13 - 14 1926 117
XM_017022737.1 XP_016878226.1 13 - 14 1607 117
XM_017022738.1 XP_016878227.1 13 - 14 1555 117
XM_017022739.1 XP_016878228.1 13 - 14 1555 117
XM_017022740.1 XP_016878229.1 14 - 15 1744 117
XM_017022741.1 XP_016878230.1 13 - 14 1957 117
XM_017022742.1 XP_016878231.1 14 - 15 1749 117
XM_017022743.1 XP_016878232.1 13 - 14 1615 117
XM_017022744.1 XP_016878233.1 13 - 14 1556 117
XM_017022745.1 XP_016878234.1 14 - 15 1744 117
XM_017022746.1 XP_016878235.1 13 - 14 1615 117
XM_017022747.1 XP_016878236.1 13 - 14 1555 117
XM_017022748.1 XP_016878237.1 13 - 14 1957 117
XM_017022749.1 XP_016878238.1 14 - 15 1717 117
XM_017022750.1 XP_016878239.1 13 - 14 1558 117
XM_017022751.1 XP_016878240.1 13 - 14 1511 117
XM_017022752.1 XP_016878241.1 14 - 15 1641 117
XM_017022753.1 XP_016878242.1 13 - 14 1615 117
XM_017022754.1 XP_016878243.1 14 - 15 1749 117
XM_017022755.1 XP_016878244.1 13 - 14 1615 117
XM_017022756.1 XP_016878245.1 13 - 14 1553 117
XM_017022757.1 XP_016878246.1 13 - 14 1511 117
XM_017022758.1 XP_016878247.1 8 - 9 901 117
XM_017022759.1 XP_016878248.1 13 - 14 1555 117
XM_017022760.1 XP_016878249.1 13 - 14 1615 117
GenBank mRNA AB002375.1 - 13 - 14 1563 117
AF502586.1 - 13 - 14 1531 117
AF502587.1 - 13 - 14 1563 117
AF502588.1 - 13 - 14 1531 117
AK307438.1 - 14 - 15 1605 117
BC050263.1 - 12 - 13 1587 117
BC057395.1 - 13 - 14 1587 117
BX647814.1 - 14 - 15 1596 117

View Full Product Details