Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PPIP5K1
- Entrez Gene ID
-
9677
- Gene Name
- diphosphoinositol pentakisphosphate kinase 1
- Gene Aliases
-
HISPPD2A, IP6K, IPS1, VIP1, hsVIP1
- Chromosome Location
-
Chr.15: 43533462 - 43590272 on Build GRCh38
View Full Product Details