The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001182.4 | NP_001173.2 | 17 - 18 | 1747 | 74 |
NM_001201377.1 | NP_001188306.1 | 17 - 18 | 1747 | 74 | |
NM_001202404.1 | NP_001189333.2 | 15 - 16 | 1555 | 74 | |
XM_011543417.2 | XP_011541719.1 | 17 - 18 | 1738 | 74 | |
XM_017009493.1 | XP_016864982.1 | 17 - 18 | 1523 | 74 | |
GenBank mRNA | AK092507.1 | - | 8 - 9 | 1759 | 74 |
AK295526.1 | - | 12 - 13 | 982 | 74 | |
AK297365.1 | - | 15 - 16 | 1501 | 74 | |
AK312459.1 | - | 17 - 18 | 1576 | 74 | |
BC002515.2 | - | 17 - 18 | 1564 | 74 | |
BC071712.1 | - | 17 - 18 | 1541 | 74 | |
BC073174.1 | - | 17 - 18 | 1533 | 74 | |
S74728.1 | - | 17 - 18 | 1525 | 74 |