Product Details

Assay Reference Genome
Location

Chr.19:18783077 on build GRCh38
Cytoband
19p13.11
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1311
Gene Symbol
COMP
Gene Name
cartilage oligomeric matrix protein
Gene Aliases
EDM1, EPD1, MED, PSACH, THBS5, TSP5
Location
Chr.19:18782773-18791305 on build GRCh38
Assay Gene Location
Overlaps Intron 18 - Exon 18
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
COMP NM_000095.2 NP_000086.2
AB086984.1 BAC53888.1
AK074508.1 BAC11031.1
AK223216.1 BAD96936.1
AK290595.1
AK296586.1
AK297798.1
AK297814.1
BC110847.1 AAI10848.1
BC125092.1
L32137.1 AAA57253.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1151217 Chr19:18627369 - 19284914 on Build GRCh38 Deletion RFXANK UPF1 HAPLN4 SUGP1 CRTC1 HOMER3 GDF1 SLC25A42 COMP DDX49 ARMC6 NR2C2AP MEF2B TMEM161A SUGP2 NCAN COPE CERS1 LOC102724360 TM6SF2 BORCS8 BORCS8-MEF2B KLHL26
nsv952422 Chr19:18606691 - 18807891 on Build GRCh38 Deletion TMEM59L CRTC1 CRLF1 KLHL26 COMP
nsv833777 Chr19:18765126 - 18920267 on Build GRCh38 Loss UPF1 COPE CERS1 CRTC1 GDF1 COMP DDX49

View Full Product Details