The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286838.1 | NP_001273767.1 | 15 - 15 | 2960 | 136 |
NM_001510.3 | NP_001501.2 | 16 - 16 | 3245 | 136 | |
XM_011531893.2 | XP_011530195.2 | 16 - 16 | 9482 | 136 | |
XM_011531894.2 | XP_011530196.1 | 14 - 14 | 2521 | 136 | |
XM_011531895.2 | XP_011530197.1 | 14 - 14 | 2438 | 136 | |
XM_017008118.1 | XP_016863607.1 | 15 - 15 | 9434 | 136 | |
XM_017008119.1 | XP_016863608.1 | 16 - 16 | 3019 | 136 | |
XM_017008123.1 | XP_016863612.1 | 14 - 14 | 3749 | 136 | |
XM_017008124.1 | XP_016863613.1 | 13 - 13 | 2969 | 136 | |
XM_017008125.1 | XP_016863614.1 | 13 - 13 | 2466 | 136 | |
XM_017008126.1 | XP_016863615.1 | 12 - 12 | 2182 | 136 | |
GenBank mRNA | AB209318.1 | - | 16 - 16 | 3245 | 136 |
AF009014.1 | - | 16 - 16 | 2987 | 136 | |
BC099652.3 | - | 16 - 16 | 2987 | 136 | |
BC099653.3 | - | 16 - 16 | 2987 | 136 | |
BC099654.3 | - | 15 - 15 | 2702 | 136 |