The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278622.1 | NP_001265551.1 | 4 - 4 | 680 | 143 |
NM_002873.1 | NP_002864.1 | 3 - 3 | 785 | 143 | |
NM_133338.2 | NP_579916.1 | 5 - 5 | 681 | 143 | |
NM_133339.2 | NP_579917.1 | 2 - 2 | 238 | 143 | |
NM_133340.2 | NP_579918.1 | 2 - 2 | 271 | 143 | |
NM_133342.2 | NP_579920.1 | 4 - 4 | 658 | 143 | |
NM_133343.1 | NP_579921.1 | 4 - 4 | 623 | 143 | |
NM_133344.2 | NP_579922.1 | 3 - 3 | 440 | 143 | |
XM_017009679.1 | XP_016865168.1 | 4 - 4 | 500 | 143 | |
XM_017009680.1 | XP_016865169.1 | 3 - 3 | 554 | 143 | |
XM_017009681.1 | XP_016865170.1 | 3 - 3 | 412 | 143 | |
GenBank mRNA | AF017748.1 | - | 3 - 3 | 410 | 143 |
AF076838.1 | - | 3 - 3 | 785 | 143 | |
AF085736.2 | - | 2 - 2 | 238 | 143 | |
AF098533.1 | - | 2 - 2 | 217 | 143 | |
AF112263.1 | - | 2 - 2 | 262 | 143 | |
AF126424.1 | - | 5 - 5 | 680 | 143 | |
AJ001642.1 | - | 3 - 3 | 449 | 143 | |
AJ004977.1 | - | 3 - 3 | 445 | 143 | |
AK292487.1 | - | 4 - 4 | 658 | 143 | |
AL122068.1 | - | 4 - 4 | 623 | 143 | |
BC032304.1 | - | 2 - 2 | 251 | 143 | |
BX537441.1 | - | 4 - 4 | 680 | 143 |