Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SCNN1D
- Entrez Gene ID
-
6339
- Gene Name
- sodium channel epithelial 1 delta subunit
- Gene Aliases
-
ENaCd, ENaCdelta, SCNED, dNaCh
- Chromosome Location
-
Chr.1: 1280415 - 1292029 on Build GRCh38
View Full Product Details