Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS28
Entrez Gene ID
6234
Gene Name
ribosomal protein S28
Gene Aliases
DBA15, S28
Chromosome Location
Chr.19: 8321500 - 8322396 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031.4 NP_001022.1 1 - 1 68 135
GenBank mRNA AK311925.1 - 1 - 2 68 135
AU126783.1 - 1 - 2 68 135
BC000354.1 - 1 - 1 54 135
BC018810.1 - 1 - 1 54 135
BC021239.2 - 1 - 1 47 135
BC070217.1 - 1 - 1 68 135
BC070218.1 - 1 - 1 65 135
CR457055.1 - 1 - 1 37 135
L05091.1 - 1 - 2 52 135
U58682.1 - 1 - 2 49 135

View Full Product Details