Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP5J2
Entrez Gene ID
9551
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit F2
Gene Aliases
ATP5JL
Chromosome Location
Chr.7: 99458161 - 99466201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003714.2 NP_001003714.1 2 - 3 206 101
NM_001039178.2 NP_001034267.1 2 - 3 188 101
GenBank mRNA CB110074.1 - 2 - 3 173 101
CD176299.1 - 2 - 3 168 101

View Full Product Details