The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000269.2 | NP_000260.1 | 5 - 5 | 726 | 121 |
NM_198175.1 | NP_937818.1 | 6 - 6 | 946 | 121 | |
GenBank mRNA | AF487339.1 | - | 6 - 6 | 930 | 121 |