Product Details

Assay Reference Genome
Location

Chr.4:89726597 on build GRCh38
Cytoband
4q22.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
6622
Gene Symbol
SNCA
Gene Name
synuclein alpha
Gene Aliases
NACP, PARK1, PARK4, PD1
Location
Chr.4:89724099-89838324 on build GRCh38
Assay Gene Location
Within Exon 10
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
SNCA NM_000345.3 6 717 NP_000336.1
NM_001146054.1 6 713 NP_001139526.1
NM_001146055.1 6 524 NP_001139527.1
NM_007308.2 5 629 NP_009292.1
XM_011532208.2 6 594 XP_011530510.1
XM_017008562.1 6 588 XP_016864051.1
XM_017008563.1 6 614 XP_016864052.1
AK290169.1 6 538
AY049786.1 AAL15443.1
BC013293.2 6 877 AAH13293.1
BC108275.1 6 671 AAI08276.1
CR457058.1 CAG33339.1
CR541653.1 CAG46454.1
D31839.1 4 383 BAA06625.1
DA391115.1 6 524
JN709859.1 5 671
JN709860.1 4 587
JN709861.1 5 633
JN709862.1 5 675
JN709863.1 4 591
JN709864.1 5 440
JN709865.1 5 482
JN709866.1 4 398
JN709867.1 5 482
JN709868.1 5 524
JN709869.1 4 440
L08850.1 6 506 AAA16117.1
L36674.1 AAA98493.1
L36675.1 AAA98487.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1012406 Chr4:89246630 - 90245636 on Build GRCh38 Loss CCSER1 GPRIN3 MMRN1 SNCA SNCA-AS1
nsv594806 Chr4:88621130 - 90539537 on Build GRCh38 Gain FAM13A TIGD2 HERC3 CCSER1 GPRIN3 FAM13A-AS1 MMRN1 SNCA SNCA-AS1 NAP1L5

View Full Product Details