Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BBS12
Entrez Gene ID
166379
Gene Name
Bardet-Biedl syndrome 12
Gene Aliases
C4orf24
Chromosome Location
Chr.4: 122700437 - 122744943 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152618.2 NP_689831.2 1 - 2 184 78
GenBank mRNA AK123553.1 - 1 - 2 184 78
BC055426.1 - 1 - 2 182 78
BX538148.1 - 1 - 2 123 78

View Full Product Details