Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TADA1
Entrez Gene ID
117143
Gene Name
transcriptional adaptor 1
Gene Aliases
ADA1, HFI1, STAF42, TADA1L, hADA1
Chromosome Location
Chr.1: 166856510 - 166876417 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053053.3 NP_444281.1 2 - 3 347 102
GenBank mRNA AK290964.1 - 2 - 3 164 102
AK291922.1 - 2 - 3 287 102
AK309250.1 - 2 - 3 215 102
BC015401.2 - 2 - 3 172 102
CN399642.1 - 2 - 3 349 102

View Full Product Details