Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKD1L2
Entrez Gene ID
114780
Gene Name
polycystin 1 like 2 (gene/pseudogene)
Gene Aliases
PC1L2
Chromosome Location
Chr.16: 81100879 - 81220370 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076780.1 NP_001070248.1 5 - 6 908 76
NM_052892.3 NP_443124.3 5 - 6 908 76
NR_126532.1 5 - 6 908 76

View Full Product Details